Tlačiť
Measuring scoops
Dodávateľ: VITLAB
39194 39294 39394 39494 39594 39694 39794 39894 39994
231-0326EA 6.18 EUR
231-0326 231-0327 231-2000 231-2001 231-2002 231-2003 231-2004 231-0218 231-2005
Measuring scoops
Lopatky
PP, white. With flat, chamfered ends and reinforced handle.
  • Excellent chemical resistance, robust
  • Heat resistant up to +130 °C, autoclavable
  • The capacity of the scoop is stamped into the handle for easy reading