Tlačiť
Scoops
Dodávateľ: RSG
758.050 758.075 758.100 758.120 758.150
231-0201EA 28.28 EUR
231-0201 231-0202 231-0203 231-0204 231-0236
Scoops
Lopatky
Stainless steel. Manual scoops for collecting or measuring powders, pastes or granulates.
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance
  • Robust design