Tlačiť
Measuring scoops, Azlon®
Dodávateľ: DWK LIFE SCIENCES
SWL030 SWL018 SWL008 SWL012 SWL036 SWL140 SWL024
231-0025EA 66.49 EUR
231-0025 231-0024 231-0022 231-0023 231-0026 231-0027 231-0472
Measuring scoops, Azlon®
Lopatky
PP, white. Scoops with flat, bevelled end and reinforced handle.
  • Excellent chemical resistance, robust
  • Heat resistant up to +130 °C, autoclavable
  • The capacity of the scoop is stamped into the handle for easy reading