Tlačiť
Measuring scoops
Dodávateľ: KARTELL
1059 1058 1060 1062 1057 1063 1064
KART1059EA 61.77 EUR
KART1059 231-0067 231-0068 231-0069 231-0066 231-0070 231-0071
Measuring scoops
Lopatky
PP, white. With flat, bevelled end and reinforced handle.
  • Excellent chemical resistance, unbreakable
  • Heat resistant up to 130 °C, autoclavable
  • The capacity of the scoop is stamped into the handle for easy reading