Tlačiť
PowderThief Single Use Samplers
Dodávateľ: SAMPLING SYSTEMS
8060H-1010 8060H-1010S 8060H-510 8060H-510S 8060H-520 8060H-520S 8060H-1001 8060H-1001S 8060H-1005 8060H-1005S 8060H-1020 8060H-1020S 8060H-501 8060H-501S 8060H-505 8060H-505S 8060H-1520 8060H-1520S 8060H-502S 8060H-503S 8060H-1000.5S 8060H-1003S
300-2016EA 491.78 EUR
300-2016 300-2017 300-2018 300-2019 300-2020 300-2021 300-2022 300-2023 300-2024 300-2025 300-2026 300-2027 300-2028 300-2029 300-2030 300-2031 300-0530 300-0531 SAMP8060H-502S SAMP8060H-503S SAMP8060H-1000.5S SAMP8060H-1003S
PowderThief Single Use Samplers
Vzorkovače Vzorkovače jednorázové
Disposable samplers specially designed for powders and granules.
  • No risk of cross-contamination
  • Assembled in a cleanroom environment
  • Economical - Save both time and money
Suitable for industries such as pharmaceutical, food, chemical, cosmetic, biotechnology. Pre-sterilised versions are available.
Certifikácie: FDA conformity. BSE/TSE free. Full batch traceability, ATEX rated.
Balenie: Each sampler is wrapped in a plastic bag.