Tlačiť
Stand plates
Dodávateľ: USBECK LABORGERATE
2000 2002 2003 2004 2005
241-7110EA 20.75 EUR
241-7110 241-7111 241-7176 241-7112 241-7113
Stand plates
Stojany laboratorní
Steel, powder coated. Firm, non slip stand. For a wide variety of uses. With M10 threaded hole on face side.
  • Rubber feet for a firm footing
  • Easy to clean
  • Robust