Tlačiť
Tripod stands
Dodávateľ: USBECK LABORGERATE
2021 2022 2023 2020
241-0245EA 29.48 EUR
241-0245 241-0246 241-0341 241-0377
Tripod stands
Stojany laboratorní
Steel, powder coated.
  • With M10 threaded hole
  • No support rod
  • Non-slip, rubber feet ensure firm footing