Tlačiť
Retort rings with bosshead
Dodávateľ: USBECK LABORGERATE
2930 2931 2932
241-0328EA 16.58 EUR
241-0328 241-0329 241-0330
Retort rings with bosshead
Kruhy ke stojanům
Galvanised steel (8 mm Ø).
  • With integral bosshead for fixing to retort stand
  • With three-point bearing
  • Nickel plated brass screw