Tlačiť
Retort rings without bosshead
Dodávateľ: USBECK LABORGERATE
2900 2901 2902 2910 2911 2912
241-0375EA 9.97 EUR
241-0375 241-0376 241-0383 241-0221 241-0222 241-0223
Retort rings without bosshead
Kruhy ke stojanům
Steel, zinc plated.
  • With three-point bearing