Tlačiť
Retort rings with bosshead
Dodávateľ: USBECK LABORGERATE
2920 2921 2922
241-7525EA 16.7 EUR
241-7525 241-7526 241-7527
Retort rings with bosshead
Kruhy ke stojanům
Galvanised steel (8 mm Ø).
  • With integral bosshead for fixing to retort stand
  • Three-point bearing
  • Nickel plated brass screw