Tlačiť
Clamp locks for attaching reaction vessels to retort stands
Dodávateľ: LENZ LABORGLAS
06246510 06246515 06246506
201-1140EA 636.98 EUR
201-1140 201-1141 201-1139
Clamp locks for attaching reaction vessels to retort stands
Svorky Svorky pro běžné účely
Fibreglass-reinforced polyester.
  • Fast, easy assembly and disassembly
  • Very sturdy
  • Secure fixture holder on the reaction vessel
For securing a reaction vessel with a fixture using an attached rod onto a retort stand.