Tlačiť
Test tube racks for water baths, SP Bel-Art
Dodávateľ: Bel-Art Products, a Part of SP
F188670001 F188640001
212-6255EA 132.99 EUR
212-6255 212-6258
Test tube racks for water baths, SP Bel-Art
Stojany Tube Racks
PP, white.
  • Excellent chemical resistance
  • Heat resistant up to +121 °C, autoclavable
  • Very stable
Due to its special blend of materials, this rack can be submerged without additional weight on the base of the bath.