Tlačiť
Test tube racks, Nalgene®
Dodávateľ: Thermo Fisher Scientific
5929-0016 5929-0020 5929-0030 5930-0016 5930-0020 5930-0030
212-8322EA 115.52 EUR
212-8322 212-8323 212-8324 212-8332 212-8333 212-8335
Test tube racks, Nalgene®
Stojany Tube Racks
PC, clear with white bottom plate or PP, white.
  • PC: autoclavable (will reduce strength over time), do not float in water baths
  • Good chemical resistance
  • PP: not autoclavable
  • Excellent chemical resistance
  • All racks are provided with alphanumeric position markings for easier sample identification
Ergonomic handles for easier transport of full racks. Empty racks can be stacked with empty racks of the same length.