Tlačiť
Microtube and test tube racks, floating
Dodávateľ: HEATHROW
HS2165C HS2165B HS2165A
212-9125EA 25.15 EUR
212-9125 212-9126 212-9127
Microtube and test tube racks, floating
Stojany Tube Racks
Polyfoam, coloured.
  • These stands float and remain on the surface of the bath, preventing the vessels they contain from floating free
  • A variety of geometrical shapes enable individual and easily sortable arrangement in the water bath
  • With handles for easy insertion and removal
For incubating sample, centrifuge and microcentrifuge tubes in water baths.