Tlačiť
Test tube racks
Dodávateľ: KARTELL
370 559 560 562
212-6100EA 21.66 EUR
212-6100 212-1420 212-1421 212-1422
Test tube racks
Stojany Tube Racks
PP, white.
  • Excellent chemical resistance
  • Heat resistant up to 121 °C, autoclavable
  • Light, very robust
With two or three tiers.