Tlačiť
Test tube racks, Unwire™, Half-Racks, Nalgene®
Dodávateľ: Thermo Fisher Scientific
5972-0013 5972-0016 5972-0020 5972-0025 5972-0030 5972-0313 5972-0316 5972-0320 5972-0325 5972-0330 5972-0413 5972-0416 5972-0420 5972-0430 5972-0513 5972-0516 5972-0520 5972-0530
212-8211EA 29.56 EUR
212-8211 212-8221 212-8231 212-8241 212-8251 212-8214 212-8224 212-8234 212-8244 212-8254 212-8215 212-8225 212-8235 212-8255 212-8216 212-8226 212-8236 212-8256
Test tube racks, Unwire™, Half-Racks, Nalgene®
Stojany Tube Racks
ResMer™, colour coded.
  • Heat resistant up to +135 °C, autoclavable
  • Half size, so more space saving than conventional racks
  • Good chemical resistance
The ResMer™ manufacturing technology improves the shelf life of these test tube racks under the harshest conditions such as autoclaving, cleaning in dishwashers, chemical disinfection and use in freezing conditions. They are unmistakeable with durable and brighter colours. The racks do not float, so they are also very suitable for use in ice or water baths.