Tlačiť
Microtube and cryotube racks
Dodávateľ: SIMPORT SCIENTIFIC
T315-3 T315-2
212-9561EA 66.99 EUR
212-9561 212-9610
Microtube and cryotube racks
Stojany Tube Racks
PP, coloured.
  • Special locking system on the base holds tubes fast, so that they do not turn when the screw cap is opened or closed
  • 50 holes (5×10) with alphanumeric labelling for sample identification
  • five anti-skid feet, stackable
For reaction vessels or cryotubes with screw caps, autoclavable.