Tlačiť
Racks for cryoboxes, Nalgene®
Dodávateľ: Thermo Fisher Scientific
5036-0004 5036-0009 5038-4322 5038-4422 DS5035-0004 DS5035-0009 DS5037-0004 DS5037-0007
212-8117EA 195.71 EUR
212-8117 212-8118 212-8119 212-8120 479-3351 479-3352 479-3356 479-3357
Racks for cryoboxes, Nalgene®
Stojany Poličky pro kryoboxy
Stainless steel.
Retainers hold each box securely but separately and allow fast retrieval of one box and immediate return to rack.
Vertical
479-3351, 479-3352: for cryoboxes with 5×5 cryotubes
212-8117, 212-8118: for standard cryoboxes
479-3356, 479-3357: for cryoboxes with 5 ml tubes and Mr. Frosty
Horizontal
Fit most upright freezer compartments
For standard cryoboxes of 133×133×51 mm
With two lateral handles
Informace o dodávce: Supplied without cryoboxes.