Tlačiť
Horizontal developing chambers
Dodávateľ: CAMAG
022.8530 022.8535
552-0047EA 1425.59 EUR
552-0047 552-0048
Horizontal developing chambers
TLC vyvíjecí komory
Horizontal developing chambers allows the number of samples to be doubled when compared to a vertical tank. Plates are developed from opposite sides simultaneously towards the middle.
  • Choice of two configurations: sandwich or tank
  • Suitable for all types of solvents
  • Available in two plates sizes