Tlačiť
PLC plates, silica gel 60, unmodified and modified layers, glass backed
Dodávateľ: Merck
1.05434.0001 1.05744.0001 1.05745.0001 1.13792.0001 1.13793.0001 1.13794.0001 1.13894.0001 1.13895.0001
1.05434.0001EA 1685.92 EUR
1.05434.0001 1.05744.0001 1.05745.0001 1.13792.0001 1.13793.0001 1.13794.0001 1.13894.0001 1.13895.0001
PLC plates, silica gel 60, unmodified and modified layers, glass backed
TLC desky
PLC (Preparative Layer Chromatography) plates are based on the same proven Merck silica-binder technology as analytical TLC plates. Plates are available with layers of unmodified silica gel or RP-18 modified silica gel in several layer thicknesses, ranging from 500 to 2000 µm, with or without fluorescent indicator.
  • Enables high sample loading
  • Separation and purification of mg up to g samples