Tlačiť
Axygen® Automation Tips for PerkinElmer, Zymark® RapidPlate®, Sciclone®
Dodávateľ: Corning
ZT-100-R ZT-100-R-S ZT-200-R ZT-200-R-S ZT-205-WBRS ZT-50-R ZTF-100-R-S ZT-50-R-S ZTF-200-R-S ZTF-50-R-S
613-1666EA 401.12 EUR
613-1666 613-1667 613-1668 613-1669 613-1671 613-1905 613-1672 613-1906 613-1673 732-1555
Axygen® Automation Tips for PerkinElmer, Zymark® RapidPlate®, Sciclone®
Špičky pipetovací
Transparent PP.
  • Sterile tips: Sterilised by gamma radiation
  • Filter tips: PE filter prevents any pipette contamination
  • Free of RNase, DNase, DNA and pyrogens
Available options:
- With or without aerosol-resistant filters
- Nonsterile or sterile
- Standard or Axygen® Maxymum Recovery® surface