Tlačiť
Robotic tips, Axygen®, for Beckman®
Dodávateľ: Corning
FXF-384XL-RS FXF-50-R-S FX-20-R FX-250-R FXF-165-WB-R-S FX-10-R FX-50-R FX-20-R-S FX-50-R-S FX-250-R-S FXF-20-R-S FX-255-WB-R FXF-180-R-S FXF-200-R-S FX-384-R FX-384-R-S FX-384-XL-R FX-384-XL-RS FXF-384-LRS FXF-205-WB-R-S
736-0275EA 1453.23 EUR
736-0275 736-0254 AXYGFX-20-R AXYGFX-250-R AXYGFXF-165-WB-R-S AXYGFX-10-R AXYGFX-50-R AXYGFX-20-R-S AXYGFX-50-R-S AXYGFX-250-R-S AXYGFXF-20-R-S AXYGFX-255-WB-R AXYGFXF-180-R-S AXYGFXF-200-R-S AXYGFX-384-R AXYGFX-384-R-S AXYGFX-384-XL-R AXYGFX-384-XL-RS AXYGFXF-384-LRS AXYGFXF-205-WB-R-S
Robotic tips, Axygen®, for Beckman®
Špičky pipetovací
Free of RNase, DNase, DNA and pyrogens.
  • Transparent PP
  • Sterile tips: Sterilised by gamma radiation
  • Filter tips: PE filter prevents any contamination of the pipette
  • Maxymum Recovery™ tips: Ultra-smooth surface prevents retention, loss of components, and denaturation of the sample