Tlačiť
Pipette tips, Finntip™ Stepper
Dodávateľ: Thermo Fisher Scientific
9404170 9404173 9404180 9404183 9404190 9404193 9404200 9404203 9404210 9404213 9404220 9404223 9404230 9404233
613-2634EA 162.45 EUR
613-2634 613-2635 613-2636 613-2637 613-2638 613-2639 613-2640 613-2641 613-2642 613-2643 613-2644 613-2645 613-2646 613-2647
Pipette tips, Finntip™ Stepper
Špičky pipetovací
Full range of positive displacement syringe tips designed for the Finnpipette™ Stepper.
  • Ergonomic design
  • Seven syringe sizes covering a range of dispensing volumes from 10 µl to 50 ml
  • Adapted for applications using aggressive and viscous liquids