Tlačiť
Special pipette tips, Finntip™
Dodávateľ: Thermo Fisher Scientific
9400100 9400130 9400133 9401410 9405020 9405050 9405120 9405123 9405160 9401420 9401423 9405163
613-2591EA 60.65 EUR
613-2591 613-2592 613-2593 613-2620 613-2648 613-2650 613-2652 613-2653 613-4488 613-4486 613-4487 613-4489
Special pipette tips, Finntip™
Špičky pipetovací
High quality PP, autoclavable.
  • Finntip™ extended (200 and 1000 µl): long, tapered tip especially adapted for deep tubes and narrow necked bottles
  • Finntip™ wide: tips with a wide, ground orifice for pipetting viscous liquids and fragile suspensions of cells or macromolecules, such as genomic DNA, without fragmentation