Tlačiť
Pipette tips, Maxymum Recovery™, Axygen®
Dodávateľ: Corning
T-1000-C-L T-1000-C-L-R T-1000-C-LRS T-200-C-L T-200-C-L-R T-200-C-LSTK T-300-L T-300-L-R T-300-L-R-S T-300-L-STK T-300-L-STKS T-350-C-L T-350-C-L-R T-350-C-L-RS T-400-L T-400-L-R T-400-L-R-S TR-222-C-L TR-222-C-L-R TR-222-C-LRS TR-222-CL-STK TXL-10-L TXL-10-L-R TXL-10-L-R-S T-200-C-L-RS
613-0457EA 37.75 EUR
613-0457 613-0458 613-0459 613-0443 613-0444 613-0446 613-0436 613-0437 613-0438 613-0439 613-1308 613-0454 613-0455 613-0456 613-0440 613-0441 613-0442 613-0447 613-0448 613-0449 613-0450 732-7512 732-7513 732-0864 613-0665
Pipette tips, Maxymum Recovery™, Axygen®
Špičky pipetovací
PP, transparent.
  • Ultra-smooth surface avoids sample retention, saving costly reagents and precious samples
  • Tips can be fitted and ejected effortlessly thanks to their supple walls and internal sealing rings
  • Certified free of RNase, DNase and endotoxins
Objednávacie informácie: Available loose, in racks or rack towers, sterile or non sterile.