Tlačiť
VWR® MultiTray®/MultiBox® plus, Pipette Tips
Dodávateľ: VWR Collection
612-2914EA 59.77 EUR
612-2914 612-2915 612-2599 612-2913 612-2999
VWR® MultiTray®/MultiBox® plus, Pipette Tips
Špičky pipetovací
Tips racked in MultiTrays®. Compatible with most micropipettes, except 612-2913 which is compatible with Gilson® and Labmate® micropipettes. MultiBox® plus is an autoclavable PP box with dustproof, hinged and sliding lid, which may be opened completely or partially. MultiBox® plus is suitable for use with all MultiTrays®.
  • Tips fit firmly on micropipettor cones, have straight shape for optimum handling, and offer minimum fluid retention
  • MultiBox® plus can be repeatedly autoclaved, but should be placed in an adapted container to avoid contact with the autoclave walls (sterilisation conditions: +121 °C, 1,2 bar, max. 30 minutes, with vessels open)
  • Up to a maximum of five MultiBox® plus can be stacked
Objednávacie informácie: Tips supplied pre-assembled in MultiTray® systems.