Tlačiť
Filter tips, Classic / Maxymum Recovery™, Axygen®
Dodávateľ: Corning
TF-100 TF-1000 TF-1000-L-R-S TF-1000-R-S TF-1005-WB-R-S TF-100-R-S TF-150-R-S TF-20 TF-200 TF-200-R-S TF-205WB-RS TF-20-L-R-S TF-20-R-S TF-300 TF-300-L-R-S TF-300-R-S TF-350 TF-350-L-R-S TF-350-R-S TF-400-L-R-S TF-400-R-S TF-420-L-R-S TF-420-R-S TF-50 TF-50-L-R-S TF-50-R-S TXLF-10 TXLF-10-R-S TF-200-L-R-S TF-100-L-R-S TF-1200-C-R-S TF-400 TXLF-10-L-R-S
613-3817EA 102.29 EUR
613-3817 613-3818 732-0863 732-0647 732-0648 732-0642 732-0643 613-3820 613-3821 732-0644 732-0645 732-0652 732-0640 613-3822 732-0649 732-0637 613-3823 732-0937 732-0646 732-0651 732-0639 732-0936 732-1407 613-3825 732-0653 732-0641 613-3831 732-0638 732-0655 732-0862 732-1404 613-3824 732-7511
Filter tips, Classic / Maxymum Recovery™, Axygen®
Špičky pipetovací
PP, sterile or non sterile.
  • Free of DNA, RNase, DNase, pyrogens and metals
  • Maxymum Recovery™ tips have an ultra-smooth surface designed to reduce sample retention and sample loss, and to avoid sample denaturation
  • Sterilised by gamma radiation