Tlačiť
Serological pipettes, Sterilin®
Dodávateľ: THERMO STERILIN
7077-1N 7078-5N 7077-2N 7077-5N 7079-5N 7078-10N 7077-10N 7078B-1 7078B-50 7079-10N 7079-1N 7078-1CN
391-2033EA 379.06 EUR
391-2033 391-2034 391-2036 391-2037 391-2038 391-2039 391-2040 SLIN7078B-1 SLIN7078B-50 SLIN7079-10N SLIN7079-1N SLIN7078-1CN
Serological pipettes, Sterilin®
Pipety Pipety serologické
Pyrex® borosilicate glass, disposable.
  • Material is chemical resistant
  • Easy to read graduations
  • Calibrated for delivery with the last drop expelled by blowing
  • Available either plugged or unplugged
Balenie: Bulk or individual paper-peel package.