Tlačiť
Micro capillaries, disposable, minicaps
Dodávateľ: HIRSCHMANN
9000201 9000203 9000204 9000225 9000299 9000202 9000205 9000210
720-2835EA 136.78 EUR
720-2835 720-2836 720-2837 612-2649 612-2650 720-2762 612-2447 612-2448
Micro capillaries, disposable, minicaps
Pipety Mikrokapiláry
Disposable capillary pipettes with the end-to-end system. With genuine Schott DURAN® glass. Accuracy ±≤0,5%, CV ±≤1%. With batch number on the package.
  • Correct filling of the capillary
  • Conformity certified, according to the German 'Eichordnung' certification
  • Colour-coded vial cap with capillary dispenser
  • Safe packaging for protection during transportation
Balenie: 100 minicaps packed in a glass vial with dispenser cap. 10 vials plus a pipetting device are packed into a outer carton.