Tlačiť
Micro haematocrit capillaries, disposable
Dodávateľ: HIRSCHMANN
9100418 9100160 9100175 9100260 9100275 9100299 9030208
612-3422EA 70.9 EUR
612-3422 612-3414 612-3417 612-3418 612-3419 720-2851 720-2948
Micro haematocrit capillaries, disposable
Pipety Mikrokapiláry
Disposable capillaries for centrifugation of blood tests.
  • Even and heavy wall thickness
  • With printed colour-code
According to DIN 12846
Outer Ø: 1,3/1,4 mm (60 µl), 1,5/1,6 mm (75/100 µl), 1,3/1,4 mm (18 µl)
Inner Ø: 0,9/1,0 mm (60 µl), 1,1/1,2 mm (75/100 µl), 0,5/0,6 mm (18 µl)
Certifikácie: According to DIN 12846
Balenie: 100 pieces packed in a glass vial, 10 vials in a carton.