Tlačiť
Graduated pipettes, type 3, Class B, total delivery
Dodávateľ: HIRSCHMANN
1000129 1000133 1000135 1000140 1000141 1000142 1000153 1000154 1000160 1000165 1000170 1000175
612-1067EA 9.11 EUR
612-1067 612-1068 612-1069 612-3400 612-1070 612-1095 612-1096 612-1097 612-1098 612-1075 612-1076 612-3401
Graduated pipettes, type 3, Class B, total delivery
Pipety Pipety graduované
AR-Glas®
  • Calibrated to deliver 'Ex'
  • Short line graduation, graduated to the tip, zero at the top, amber graduation
  • From 5 ml with a cotton plug at the upper end as standard
DIN EN ISO 835
Length: 360 mm ±5 mm except for 25 and 50 ml pipettes: 450 mm ±5 mm
Certifikácie: DIN EN ISO 835