Tlačiť
Double-bucket carts, Slim T™
Dodávateľ: MICRONOVA
C-7 C-7WH
129-0063EA 2505.82 EUR
129-0063 141-0999
Double-bucket carts, Slim T™
Mopy
Simple and effective double-bucket system has a stainless steel frame with two 22,7 L polypropylene buckets (included).
  • The entire unit can be autoclaved, including the buckets (C-7WH model only)
  • 483 mm wide buckets makes cart suitable for use with a variety of mop heads
  • Excellent manoeuvrability on wheels that rotate 360°
  • Optional stainless steel Slim T™ wringer to wring out SlimLine, T-Mop, and other flat-pad mops