Tlačiť
Slide mailers
Dodávateľ: KARTELL
925 923 924
631-9145EA 43.32 EUR
631-9145 631-9160 631-9161
Slide mailers
Obaly na mikroskopická sklíčka
These mailers are suitable for the safe transportation of cytological and geological microscope slides.
  • Separate compartment with guide rails and more space for each slide
  • Flush closing
  • Lightweight and flat
W×D×H: 50×100×6 mm (for 1); 67×85×6 mm (for 2); 84×100×6 mm (for 3)