Tlačiť
Cover glasses, Nunc Thermanox
Dodávateľ: Thermo Fisher Scientific
150067 174934 174942
734-2154EA 873.22 EUR
734-2154 734-2115 734-2116
Cover glasses, Nunc Thermanox
Sklíčka krycí
These cover glasses remain flat during use.
  • Resistant to common solvents
  • Autofluorescent from 380 to 545 nm
  • Surfaced treatment on one side for optimal cell attachment and growth
Thickness: 0,2 mm