Tlačiť
Jednorázové robotické zásobníky, Nalgene®
Dodávateľ: Thermo Fisher Scientific
1200-1300 1200-1301 1200-2300 1200-2301
216-4371EA 433.21 EUR
216-4371 216-4372 216-4373 216-4374
Jednorázové robotické zásobníky, Nalgene®
Zásobníky na reagencie
Made from PP. A standard microplate format with 300 ml volume designed to increase assay efficiency.
  • Convoluted design minimises dead volume and incorporates baffles to reduce splashing
  • Flat bottom plate is designed to meet a variety of assay needs in both manual and automated environments
  • Sterile version is free from RNase, DNase and pyrogens
L×W×H: 86×128×43 mm
L×W×H: 86×128×43 mm