Tlačiť
PCR plates, 96-well, semi-skirted, Thermo-Fast®
Dodávateľ: Thermo Fisher Scientific
AB-0900 AB-0900/W AB-0900/B AB-0900/G AB-0900/P AB-0900/R AB-0900/Y
732-4904EA 164.46 EUR
732-4904 732-0400 732-0396 732-0397 732-0082 732-0399 732-0401
PCR plates, 96-well, semi-skirted, Thermo-Fast®
Mikrodestičky Destičky pro PCR
PP. Compatible with all major thermal cyclers.
  • Greater rigidity than non-skirted plates and provides space for barcoding
  • Segmented design allows it to be cut into 24 and 48 well sections
  • Maximum well volume 0,3 ml when used with adhesive and heat seals
  • Cut corner (A12)