Tlačiť
PCR plates, 96-well, fully skirted, low profile, Thermo-Fast®
Dodávateľ: Thermo Fisher Scientific
AB-0800 AB-0800/R AB-0800/K AB-0800/W AB-0800/B AB-0800/G AB-0800/P AB-0800/Y AB-0800/L AB-0800/Y-L AB-0800/W-L AB-0800/R-L AB-0800/P-L AB-0800/G-L AB-0800/150 AB-0800/B-L
732-4888EA 168.47 EUR
732-4888 732-0390 732-0388 732-0391 732-0078 732-0079 732-0389 732-0392 732-0881 732-2949 732-2948 732-2947 732-2946 732-2945 ADVAAB-0800/150 732-2944
PCR plates, 96-well, fully skirted, low profile, Thermo-Fast®
Mikrodestičky Destičky pro PCR
PP. The low profile of the plate decreases ‘dead space' between the heated lid of the thermal cycler and the sample. This eliminates condensation on the side-wall of the tube, preventing reduction in PCR volume and increasing the efficiency of the reaction.
  • Suitable for use with automated systems
  • Maximum well volume 0,2 ml when used with adhesive and heat seals
  • Eight holes in skirt aid plate positioning and removal from thermal cycler block
  • Clear well bottom for sample visibility
  • Certified free from DNase, RNase and human genomic DNA