Tlačiť
Cap sealing mats for 96-well storage plates
Dodávateľ: THERMO MATRIX TECHNOLOGIES
AB-0566 AB-0674 AB-0675 AB-0566/SP
732-4822EA 225.03 EUR
732-4822 732-4846 732-4847 ADVAAB-0566/SP
Cap sealing mats for 96-well storage plates
Těsnění Těsnění mikrodestičky
Round or square capped mats for sealing 96-well storage plates.
  • Alphanumeric grid-referencing to aid sample identification
  • Autoclavable or pierceable formats
  • Pierceable mats are solvent resistant (including DMSO)
  • Flexible design
Colour: Clear