Tlačiť
DeepWell™ plates, 96-well, 1,0 ml, Nunc™
Dodávateľ: Thermo Fisher Scientific
278605 278606
736-0602EA 235.53 EUR
736-0602 736-0603
DeepWell™ plates, 96-well, 1,0 ml, Nunc™
Mikrodestičky Destičky s hlubokými jamkami
PS. Ideal for sample collection, storage and combinatorial chemistry library applications.
  • Cylindrical wells with round bottom for optimal mixing and recovery
  • Easy to use with automatic sample handling instruments
  • Alphanumeric grid for quick sample identification
  • Sealable with well caps and sealing tape
  • Sterile format certified RNase- and DNase-free
Colour: Clear