Tlačiť
Destičky pro skladování hlubokých jamek, 96 jamek, 2,2 ml
Dodávateľ: THERMO MATRIX TECHNOLOGIES
AB-0661 AB-0788
732-4843EA 367.43 EUR
732-4843 732-4884
Destičky pro skladování hlubokých jamek, 96 jamek, 2,2 ml
Mikrodestičky Destičky s hlubokými jamkami
96-well PP plates with square wells for storage and screening applications, as well as growth of bacterial cultures when used in conjuction with gas permeable adhesive seals.
  • 2,2 ml working volume when sealed with adhesive or heat seals or 1,8 ml when sealed with 96-cap sealing mat
  • U-shaped well bottom to aid resuspension