Tlačiť
Protein coated cell culture plates, CELLCOAT®, collagen type I
Dodávateľ: Greiner Bio-One
655950_20 781956_20 655956_20 781950_20 657950_50 662950_50
392-0029EA 496.99 EUR
392-0029 737-0081 392-0030 392-0136 392-0272 392-0278
Protein coated cell culture plates, CELLCOAT®, collagen type I
Mikrodestičky Destičky pro bunečné kultury
PS, coated with rat tail collagen type I. Collagen type I is an extracellular matrix protein, which in vivo influences adhesion, migration and proliferation, amongst other processes. Many otherwise difficult-to-cultivate cells adhere to collagen type I and show positive growth behaviour. For certain cell lines collagen type I also has an influence on differentiation and morphology.
  • Improved cell adhesion, proliferation and growth of endothelial cells, hepatocytes, muscle cells, pheochromocytoma cells (PC12), as well as other cell types
  • Suitable for serum-free or serum-reduced cell cultivation
  • Quality control tested to demonstrate promotion of the adhesion and proliferation of human fibrocarcoma cells
Shelf life: 24 months at room temperature