Tlačiť
Cell culture plates, 96-well, CELLSTAR®
Dodávateľ: Greiner Bio-One
655098_32 655083_32 655086_32 655073_40 655088_40 655079_40 655087_40 655090_32 675083_32 675086_32 675090_32 675180_32
738-0024EA 525.87 EUR
738-0024 392-0024 392-0025 392-0022 738-0019 392-0023 738-0018 736-0230 392-0056 392-0057 392-0058 392-0060
Cell culture plates, 96-well, CELLSTAR®
Mikrodestičky Destičky pro bunečné kultury
PS, cell culture-treated, sterile. All plates are certified free of RNase, DNase and human DNA, are non-pyrogenic and non-cytotoxic.
  • F-bottom wells with excellent optical properties are ideal for microscopic applications
  • Clear bottom plates have pigmented walls and a transparent thin film bottom, ideal of fluorescence or luminescence detection methods
  • Half area plates allow a reduction of sample volume by up to 50%
  • Lid enables gas exchange with minimal evaporation
Colour: µClear® (clear) base, or solid black or white
Colour: µClear® (clear), black or white