Tlačiť
Plates, poly-D-lysine coated, Nunc™
Dodávateľ: Thermo Fisher Scientific
152028 152029 152037 152039
734-1177EA 1221.82 EUR
734-1177 734-1178 734-1184 734-1186
Plates, poly-D-lysine coated, Nunc™
Mikrodestičky Destičky pro bunečné kultury
PS, ready to use plates pre-coated with poly-D-lysine, with lid.
  • Consistent growth surface from each lot ensures guaranteed performance
  • High quality surface promotes cell attachment, growth and differentiation in serum-free and serum-containing media
  • Stable at room temperature
W×L: 128×86 mm