Tlačiť
96 MicroWell™ plates, Nunclon™∆
Dodávateľ: Thermo Fisher Scientific
143761 156545 161093 163320 167008 168055 168136
734-2033EA 170.88 EUR
734-2033 732-2601 734-2073 734-2080 734-2097 732-2607 734-1192
96 MicroWell™ plates, Nunclon™∆
Mikrodestičky Destičky pro bunečné kultury
PS, high flange design, sterile. Ideal for cell culture, cloning, viral titrations and cell fusion.
  • Plate edges and lids designed to reduce evaporation
  • Raised well rims reduce risk of cross contamination
  • Lids shaped to facilitate stacking and handling
  • 96-well footprint fits standard equipment
Colour: Clear
External L×W: 128×86 mm
External L×W: 128×86 mm