Tlačiť
White microplates for luminescence applications
Dodávateľ: Thermo Fisher Scientific
7566 7567 7571 7572
735-0523EA 249.1 EUR
735-0523 735-0524 735-0525 735-0526
White microplates for luminescence applications
Mikrodestičky Mikrodestičky a stripy pro imunologii
Microlite™ microplates are densely pigmented to minimise crosstalk and to provide high reflectivity. These characteristics are ideal for intense luminescent reactions where light piping may be a concern or minimal luminescence assays where even a minute amount of light could affect results.
  • Microlite™+ surface offers minimum crosstalk and extra high reflectivity
  • Choice of 96-well or strip plate format
  • Flat bottom wells for optimum optical characteristics
Microlite™ 1+ is the ideal vessel for low signal luminescent reactions, offering medium binding with enhanced dynamic range, extra high reflectivity and minimal crosstalk.
Microlite™ 2+ is an irradiated, high binding surface with enhanced dynamic range, extra high reflectivity and minimal crosstalk for low signal luminescent reactions.