Tlačiť
Immunology plates, 384-well, MaxiSorp™, Nunc™
Dodávateľ: Thermo Fisher Scientific
460372 460518 464718
735-0035EA 426.39 EUR
735-0035 735-0036 735-0114
Immunology plates, 384-well, MaxiSorp™, Nunc™
Mikrodestičky Mikrodestičky a stripy pro imunologii
PS, with certified MaxiSorp™ surface treatment, pinchbar design. Available with a choice of clear plates for colorimetric assays and sample storage; white plates for luminescence assays; and black plates for fluorescence assays.
  • Same footprint as standard 96-well plate
  • White and black plates minimise crosstalk
  • Rounded square wells eliminate wicking
  • Flat bottom for optical reading
  • Compatible with automated readers and liquid handling systems
Working range: 10 to 100 µl per well.
Working range: 10 - 100 µl per well