Tlačiť
Plates and modules, FluoroNunc™ and LumiNunc™
Dodávateľ: Thermo Fisher Scientific
436110 436111 437111 437112 437591 437702 437796 437915 475515 475523
735-0002EA 384.28 EUR
735-0002 735-0003 735-0004 735-0005 735-0006 735-0007 735-0008 735-0012 735-0070 735-0071
Plates and modules, FluoroNunc™ and LumiNunc™
Mikrodestičky Mikrodestičky a stripy pro imunologii
PS, with MaxiSorp™ or PolySorp™ treated surface. For fluorescence and luminescence based immunoassays and binding assays. Available with a choice of certified MaxiSorp™ surface, with high affinity to molecules with mixed hydrophilic/hydrophobic domains, or, PolySorp™ surface, less polar than MaxiSorp™ and with affinity to molecules of a more hydrophobic nature.
  • Choice of framed modules or 96-well plates with either flat (F) or C-shaped wells
  • White modules/plates give maximum reflection with minimum autofluorescence and autoluminescence
  • Black modules/plates give minimum background in fluorescence reading, allow minimum back scattered light, and minimise cross talk
  • Clear modules/plates are suitable for time-resolved fluorescence
* With certificate