Tlačiť
Measuring cylinders, low form, PMP
Dodávateľ: KARTELL
1332 1333 1334 1335 1336 1337 1338 1339
612-4000EA 9.11 EUR
612-4000 612-4001 612-4002 612-4003 612-4004 612-4005 612-4006 612-4007
Measuring cylinders, low form, PMP
Válce Válce odměrné
PMP (TPX®), transparent, low form, autoclavable.
  • Pouring spout, circular base and moulded white graduations
  • Excellent chemical resistance
  • Can withstand temperatures up to +170 °C