Tlačiť
Mixing measuring cylinders, tall form, class A, borosilicate glass
Dodávateľ: HIRSCHMANN
2340194 2340160 2340170 2340175 2340180 2340186 2340190 2340193
612-3095EA 273.16 EUR
612-3095 612-1525 612-1526 612-1527 612-1528 612-1529 612-1530 612-1531
Mixing measuring cylinders, tall form, class A, borosilicate glass
Válce Válce odměrné
DURAN®, borosilicate glass, with PE stopper, class A.
  • Calibrated to contain 'ln', certificate of conformity (DE-M marking) with dated batch identification available via website: (www.hirschmannlab.de)
  • Hexagonal glass base, NS ground joint
  • Blue graduations with ring marks at major graduations
DIN EN ISO 4788
Certifikácie: DIN EN ISO 4788