Tlačiť
Measuring cylinders, low form, PP
Dodávateľ: KARTELL
814 815 816 817 820 821 822 823
612-4722EA 6.89 EUR
612-4722 612-4724 612-4726 612-4728 612-4730 612-4732 612-4734 612-4736
Measuring cylinders, low form, PP
Válce Válce odměrné
PP, translucent, low form.
  • With lip and circular base
  • Excellent chemical resistance
  • Permanent moulded graduations